Product Details

SNP ID
rs77595588
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:441365 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCCAGGACTCCAGCGACTGCGCA[C/T]GACGGCGATTGTCGCTGCACCTGCT
Phenotype
MIM: 615928
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM101B PubMed Links

Gene Details

Gene
FAM101B
Gene Name
family with sequence similarity 101 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182705.2 2508 UTR 3 NP_874364.1

View Full Product Details