Product Details

SNP ID
rs35545523
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:10075441 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGAGGTCGTGACCAGCGTGTGCTC[A/G]TCCAGCCAGGCCAGGCTGCTGACAT
Phenotype
MIM: 604734
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR3138 PubMed Links

Gene Details

Gene
MIR3138
Gene Name
microRNA 3138
There are no transcripts associated with this gene.

Gene
WDR1
Gene Name
WD repeat domain 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005112.4 2234 Silent Mutation GAC,GAT D446D NP_005103.2
NM_017491.3 2234 Silent Mutation GAC,GAT D586D NP_059830.1
XM_017008880.1 2234 Silent Mutation GAC,GAT D639D XP_016864369.1

View Full Product Details