Product Details

SNP ID
rs61745404
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:124542851 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAACAGGGAGGACACTTTCCCTTG[C/T]TCGAGGGGCAGGATGGAAAGGGGTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR8B12 PubMed Links

Gene Details

Gene
OR8B12
Gene Name
olfactory receptor family 8 subfamily B member 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005195.1 804 Silent Mutation GAA,GAG E268E NP_001005195.1

View Full Product Details