Product Details

SNP ID
rs61738585
Assay Type
Functionally tested
NCBI dbSNP Submissions
8
Location
Chr.1:27006457 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGCCGCAGGTGCTCCAGGGCCTC[A/G]GTGAAGTCCCCGTACAGGCTTTCGC
Phenotype
MIM: 616824
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM46B PubMed Links

Gene Details

Gene
FAM46B
Gene Name
family with sequence similarity 46 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052943.3 937 Silent Mutation ACC,ACT T255T NP_443175.2
Gene
TRNP1
Gene Name
TMF1-regulated nuclear protein 1
There are no transcripts associated with this gene.

View Full Product Details