Product Details

SNP ID
rs76681357
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:197889756 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGTGAGTACGACGTATGCTATGCC[G/T]GACAAGCTGCATTCTTTCCACATAA
Phenotype
MIM: 612477
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
IQCG PubMed Links

Gene Details

Gene
IQCG
Gene Name
IQ motif containing G
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134435.2 Intron NP_001127907.1
NM_001323027.1 Intron NP_001309956.1
NM_001323028.1 Intron NP_001309957.1
NM_001323029.1 Intron NP_001309958.1
NM_001323030.1 Intron NP_001309959.1
NM_032263.4 Intron NP_115639.1
XM_017007316.1 Intron XP_016862805.1
XM_017007317.1 Intron XP_016862806.1
Gene
LRCH3
Gene Name
leucine rich repeats and calponin homology domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032773.3 Intron NP_116162.1
XM_005269362.2 Intron XP_005269419.1
XM_005269363.2 Intron XP_005269420.1
XM_005269365.2 Intron XP_005269422.1
XM_005269367.2 Intron XP_005269424.1
XM_006713791.3 Intron XP_006713854.1
XM_006713792.3 Intron XP_006713855.1
XM_006713793.2 Intron XP_006713856.1
XM_011513242.2 Intron XP_011511544.1
XM_011513243.2 Intron XP_011511545.1
XM_017007350.1 Intron XP_016862839.1
XM_017007351.1 Intron XP_016862840.1
XM_017007352.1 Intron XP_016862841.1

View Full Product Details