Product Details

SNP ID
rs1018819390
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:102398756 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAATACACTGTACATTATATGCATG[A/G]CCTGTTTATACTATTTTCAAAAAGA
Phenotype
MIM: 606356
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TMEM123 PubMed Links

Gene Details

Gene
TMEM123
Gene Name
transmembrane protein 123
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052932.2 1159 UTR 3 NP_443164.2

View Full Product Details