Product Details

SNP ID
rs7122587
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:6622745 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGAGTCACTGGCTAGGGGTGCTGC[C/T]TGTGACATTGGGCCAGGGGCTGCCT
Phenotype
MIM: 603057 MIM: 607998
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DCHS1 PubMed Links

Gene Details

Gene
DCHS1
Gene Name
dachsous cadherin-related 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003737.3 9338 Silent Mutation CAA,CAG Q2977Q NP_003728.1
Gene
TPP1
Gene Name
tripeptidyl peptidase 1
There are no transcripts associated with this gene.

View Full Product Details