Product Details

SNP ID
rs41268073
Assay Type
Functionally tested
NCBI dbSNP Submissions
28
Location
Chr.1:40541731 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCAGGTGAGTGAGAGAAATTCAGA[A/T]GGGGCTGTGTGGAGTTGAGATCCCC
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ZNF684 PubMed Links

Gene Details

Gene
ZNF684
Gene Name
zinc finger protein 684
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152373.3 Intron NP_689586.3
XM_011540671.2 Intron XP_011538973.1
XM_011540672.2 Intron XP_011538974.1
XM_011540673.2 Intron XP_011538975.1
XM_017000290.1 Intron XP_016855779.1
XM_017000291.1 Intron XP_016855780.1
XM_017000292.1 Intron XP_016855781.1

View Full Product Details