Product Details

SNP ID
rs62175194
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:159772092 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACAAAAGAGTATTTGGGAAATTAG[G/T]GTACACAAGTTGCAAGTATATTTTG
Phenotype
MIM: 612246 MIM: 613334
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CD302 PubMed Links

Gene Details

Gene
CD302
Gene Name
CD302 molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198763.1 Intron NP_001185692.1
NM_001198764.1 Intron NP_001185693.1
NM_014880.4 Intron NP_055695.2
Gene
LY75-CD302
Gene Name
LY75-CD302 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198759.1 Intron NP_001185688.1
NM_001198760.1 Intron NP_001185689.1
Gene
MARCH7
Gene Name
membrane associated ring-CH-type finger 7
There are no transcripts associated with this gene.

View Full Product Details