Product Details

SNP ID
rs28406515
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:99516482 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCCTGCTGACACTGGATGCCCTAC[A/G]AGGGAAGCAGCTCATGGCTGGGCCA
Phenotype
MIM: 603544
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCDC85C PubMed Links

Gene Details

Gene
CCDC85C
Gene Name
coiled-coil domain containing 85C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144995.1 Intron NP_001138467.1
XM_011536705.2 Intron XP_011535007.1
XM_011536706.2 Intron XP_011535008.1
XM_011536707.2 Intron XP_011535009.1
Gene
CCNK
Gene Name
cyclin K
There are no transcripts associated with this gene.

View Full Product Details