Product Details

SNP ID
rs16970510
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:35547305 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGGTTATCTAGTCTCCCTTCCTTA[C/T]TGTGACATTTTCCTGCAGGACCTTC
Phenotype
MIM: 137216 MIM: 609169 MIM: 613585
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ATP4A PubMed Links

Gene Details

Gene
ATP4A
Gene Name
ATPase H+/K+ transporting alpha subunit
There are no transcripts associated with this gene.

Gene
GAPDHS
Gene Name
glyceraldehyde-3-phosphate dehydrogenase, spermatogenic
There are no transcripts associated with this gene.

Gene
TMEM147
Gene Name
transmembrane protein 147
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242597.1 Intron NP_001229526.1
NM_001242598.1 Intron NP_001229527.1
NM_032635.3 Intron NP_116024.1
Gene
TMEM147-AS1
Gene Name
TMEM147 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details