Product Details
- SNP ID
-
hCV25943419
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:37396237 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TATTTGGCATTAAACATTTAATGTT[A/T]GAACTACAAATTTAAAAAAATTAAA
- Phenotype
-
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
WDR70
PubMed Links
Gene Details
- Gene
- WDR70
- Gene Name
- WD repeat domain 70
There are no transcripts associated with this gene.
View Full Product Details