Product Details

SNP ID
rs61737957
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:33288741 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTAATCCCTGGGGTCTGCCTCCAG[A/G]GCTGACACAGGAGCTGAAGGACCAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BPIFB1 PubMed Links

Gene Details

Gene
BPIFB1
Gene Name
BPI fold containing family B member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033197.2 277 Missense Mutation AAG,AGG K39R NP_149974.2

View Full Product Details