Product Details

SNP ID
rs61740963
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:58369246 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCTTCGAGGGGCCTCGGCTCCTC[C/G]GGCCTCTTCTCCCGGGCCCCGGAGG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LOC105372483 PubMed Links

Gene Details

Gene
LOC105372483
Gene Name
uncharacterized LOC105372483
There are no transcripts associated with this gene.

Gene
ZNF497
Gene Name
zinc finger protein 497
There are no transcripts associated with this gene.

Gene
ZNF837
Gene Name
zinc finger protein 837
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138466.1 1076 Silent Mutation CCC,CCG P29P NP_612475.1
XM_017026242.1 1076 Silent Mutation CCC,CCG P29P XP_016881731.1

View Full Product Details