Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001202858.1 | 85 | UTR 5 | NP_001189787.1 | ||
NM_004425.3 | 85 | UTR 5 | NP_004416.2 | ||
NM_022664.2 | 85 | UTR 5 | NP_073155.2 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001271895.1 | 85 | Intron | NP_001258824.1 | ||
NM_001271896.1 | 85 | Intron | NP_001258825.1 | ||
NM_025150.4 | 85 | Intron | NP_079426.2 | ||
XM_006711555.1 | 85 | Intron | XP_006711618.1 | ||
XM_006711556.1 | 85 | Intron | XP_006711619.1 | ||
XM_017002394.1 | 85 | Intron | XP_016857883.1 | ||
XM_017002395.1 | 85 | Intron | XP_016857884.1 |