Product Details

SNP ID
rs145921744
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:31679209 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCTCCAAGAGGCATCGGTAGCAGC[A/G]CAGGTATTTGGGGAATGGAAGTGGT
Phenotype
MIM: 142620 MIM: 115441 MIM: 610433 MIM: 610434
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ABHD16A PubMed Links

Gene Details

Gene
ABHD16A
Gene Name
abhydrolase domain containing 16A
There are no transcripts associated with this gene.

Gene
CSNK2B
Gene Name
casein kinase 2 beta
There are no transcripts associated with this gene.

Gene
LY6G5B
Gene Name
lymphocyte antigen 6 complex, locus G5B
There are no transcripts associated with this gene.

Gene
LY6G5C
Gene Name
lymphocyte antigen 6 complex, locus G5C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025262.3 181 Missense Mutation CGC,TGC R61C NP_079538.3

View Full Product Details