Product Details

SNP ID
hCV25952436
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:28098659 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGTCCTGGAAGGCAGGGGCATAG[C/T]TGCAGCCAACAACACACCAGGTCAC
Phenotype
MIM: 608411
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
XPO6 PubMed Links

Gene Details

Gene
XPO6
Gene Name
exportin 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270940.1 Intron NP_001257869.1
NM_015171.3 Intron NP_055986.1

View Full Product Details