Product Details

SNP ID
rs2113526
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:30833206 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCACATTTACAAACATTAAAAATC[A/G]CTAAATGTTAAAAAATATATAAGTA
Phenotype
MIM: 605795
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TEX15 PubMed Links

Gene Details

Gene
TEX15
Gene Name
testis expressed 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031271.3 9380 UTR 3 NP_112561.2
XM_005273575.3 9380 UTR 3 XP_005273632.1
XM_005273576.3 9380 UTR 3 XP_005273633.1
XM_006716369.3 9380 UTR 3 XP_006716432.1
XM_011544588.2 9380 UTR 3 XP_011542890.1
XM_011544589.2 9380 UTR 3 XP_011542891.1
XM_011544590.2 9380 UTR 3 XP_011542892.1
XM_011544591.2 9380 UTR 3 XP_011542893.1
XM_011544592.2 9380 UTR 3 XP_011542894.1
XM_011544593.2 9380 UTR 3 XP_011542895.1
XM_011544594.2 9380 Intron XP_011542896.1

View Full Product Details