Product Details

SNP ID
rs592792
Assay Type
DME
NCBI dbSNP Submissions
46
Location
Chr.1:109669334 on Build GRCh38
Set Membership
DME Validated Inventoried
Context Sequence [VIC/FAM]
GGACTCACAAGATCACCCAGAGCAA[C/T]GCCATCCTGCGGTACATTGCCCGCA
Phenotype
MIM: 138380 MIM: 138333
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GSTM2 PubMed Links

Gene Details

Gene
GSTM2
Gene Name
glutathione S-transferase mu 2 (muscle)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000848.3 316 Silent Mutation AAC,AAT N74N NP_000839.1
NM_001142368.1 316 Silent Mutation AAC,AAT N74N NP_001135840.1
Gene
GSTM4
Gene Name
glutathione S-transferase mu 4
There are no transcripts associated with this gene.

View Full Product Details