Product Details

SNP ID
rs78649601
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:38726181 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAACAGCACGAGTGTCAACAAATG[C/G]TTTTGTTGTTGCAAAAACAGTCTCA
Phenotype
MIM: 605485
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
VPS41 PubMed Links

Gene Details

Gene
VPS41
Gene Name
VPS41, HOPS complex subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014396.3 2581 UTR 3 NP_055211.2
NM_080631.3 2581 UTR 3 NP_542198.2
XM_017011988.1 2581 UTR 3 XP_016867477.1

View Full Product Details