Product Details

SNP ID
rs61743681
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:19514822 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCATGCACCCGGTGACCATGACGTA[A/G]AGCACGACGCCCATGCTCCACACAT
Phenotype
MIM: 614997 MIM: 609435 MIM: 610712
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GATAD2A PubMed Links

Gene Details

Gene
GATAD2A
Gene Name
GATA zinc finger domain containing 2A
There are no transcripts associated with this gene.

Gene
NDUFA13
Gene Name
NADH:ubiquinone oxidoreductase subunit A13
There are no transcripts associated with this gene.

Gene
TSSK6
Gene Name
testis specific serine kinase 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032037.3 839 Silent Mutation CTC,CTT L202L NP_114426.1

View Full Product Details