Product Details

SNP ID
rs34787462
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:74927255 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTCTGGGTCTCTGCTAAAGACTG[A/C]GTTCACGTCCACCTGGATCTGACCT
Phenotype
MIM: 607827 MIM: 607828 MIM: 607696
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
OTOP2 PubMed Links

Gene Details

Gene
OTOP2
Gene Name
otopetrin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178160.2 575 Silent Mutation TGA,TGC *161C NP_835454.1
XM_011525479.1 575 Nonsense Mutation TGA,TGC *161C XP_011523781.1
Gene
OTOP3
Gene Name
otopetrin 3
There are no transcripts associated with this gene.

Gene
USH1G
Gene Name
USH1 protein network component sans
There are no transcripts associated with this gene.

View Full Product Details