Product Details

SNP ID
rs189028330
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:31489548 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGAGGCTTGATGTTGATGGGTTG[G/C]TGATTAAAGCCTCCCAAAGCCAATC
Phenotype
MIM: 182381
Polymorphism
G/C, Transversion substitution
Allele Nomenclature
Literature Links
C16orf58 PubMed Links

Gene Details

Gene
C16orf58
Gene Name
chromosome 16 open reading frame 58
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022744.3 2571 UTR 3 NP_073581.2
XM_017023570.1 2571 UTR 3 XP_016879059.1
Gene
LOC105371171
Gene Name
uncharacterized LOC105371171
There are no transcripts associated with this gene.

Gene
SLC5A2
Gene Name
solute carrier family 5 member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003041.3 2571 Intron NP_003032.1
XM_006721072.3 2571 Intron XP_006721135.2

View Full Product Details