Product Details

SNP ID
rs56148171
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:96345861 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGATTGATTAAGAAGCAGCTTGGC[A/C]TCTTTCCTCTCCAAGTACAAAACCC
Phenotype
MIM: 187410
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
DNTT PubMed Links

Gene Details

Gene
DNTT
Gene Name
DNA nucleotidylexotransferase
There are no transcripts associated with this gene.

Gene
OPALIN
Gene Name
oligodendrocytic myelin paranodal and inner loop protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040102.2 685 UTR 3 NP_001035191.1
NM_001040103.2 685 UTR 3 NP_001035192.1
NM_001284320.1 685 UTR 3 NP_001271249.1
NM_001284321.1 685 UTR 3 NP_001271250.1
NM_001284322.1 685 UTR 3 NP_001271251.1
NM_001284323.1 685 UTR 3 NP_001271252.1
NM_001284324.1 685 UTR 3 NP_001271253.1
NM_001284326.1 685 UTR 3 NP_001271255.1
NM_001284327.1 685 UTR 3 NP_001271256.1
NM_033207.4 685 UTR 3 NP_149984.1

View Full Product Details