Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001172303.2 | Intron | NP_001165774.1 | |||
NM_001172304.2 | Intron | NP_001165775.1 | |||
NM_001320756.1 | Intron | NP_001307685.1 | |||
NM_001320757.1 | Intron | NP_001307686.1 | |||
NM_032844.4 | Intron | NP_116233.2 | |||
XM_005252631.3 | Intron | XP_005252688.1 | |||
XM_005252632.3 | Intron | XP_005252689.1 | |||
XM_006717519.3 | Intron | XP_006717582.1 | |||
XM_006717520.3 | Intron | XP_006717583.1 | |||
XM_011519751.2 | Intron | XP_011518053.1 | |||
XM_017016852.1 | Intron | XP_016872341.1 | |||
XM_017016853.1 | Intron | XP_016872342.1 | |||
XM_017016854.1 | Intron | XP_016872343.1 | |||
XM_017016855.1 | Intron | XP_016872344.1 | |||
XM_017016856.1 | Intron | XP_016872345.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001253866.1 | Intron | NP_001240795.1 | |||
NM_014263.3 | Intron | NP_055078.1 | |||
NM_139312.2 | Intron | NP_647473.1 |