Product Details

SNP ID
rs7937809
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:124185859 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTGCTGACACATCCTTAGCCCAGA[A/G]GGTGAGCTTCACCAACGTTGGCCTC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR10D3 PubMed Links

Gene Details

Gene
OR10D3
Gene Name
olfactory receptor family 10 subfamily D member 3 (putative)
There are no transcripts associated with this gene.

View Full Product Details