Product Details

SNP ID
hCV25953901
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:23693359 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGCAAAATAAAACCCAACAAAAAC[G/T]TATGAGGGGTGTGAATAGCACTCCC
Phenotype
MIM: 601673
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ELAVL2 PubMed Links

Gene Details

Gene
ELAVL2
Gene Name
ELAV like neuron-specific RNA binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171195.1 Intron NP_001164666.1
NM_001171197.1 Intron NP_001164668.1

View Full Product Details