Product Details

SNP ID
rs189803914
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:86753744 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATGGGGGGTGTGCAATATGAGAAA[A/G]TGGTTGGAGCTTATTTCACTTATCA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RMND5A PubMed Links

Gene Details

Gene
RMND5A
Gene Name
required for meiotic nuclear division 5 homolog A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022780.3 Intron NP_073617.1

View Full Product Details