Product Details
- SNP ID
-
hCV25954253
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:73207792 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGTGTTTGAAAGCTCAAACAGCTTG[G/T]CTTCTTACAGCATCGAGTTGTTTGC
- Phenotype
-
MIM: 606973
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
COG1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs71710311] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- COG1
- Gene Name
- component of oligomeric golgi complex 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_018714.2 |
2286 |
Intron |
|
|
NP_061184.1 |
- Gene
- FAM104A
- Gene Name
- family with sequence similarity 104 member A
View Full Product Details