Product Details

SNP ID
rs3810712
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:79170974 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGATTGTGAATTTAGTTTTGAACCA[C/T]CATTAGGCAAAGATAGTTTCTCTAG
Phenotype
MIM: 300903
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GPR174 PubMed Links

Gene Details

Gene
GPR174
Gene Name
G protein-coupled receptor 174
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032553.1 3 UTR 5 NP_115942.1

View Full Product Details