Product Details

SNP ID
hCV25954589
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:112233169 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTTGGATAATATCATTTTGGTAT[C/A]CCTAAAACACTGGTTTCATTAAAAA
Phenotype
MIM: 611551
Polymorphism
C/A, Transversion substitution
Allele Nomenclature
Literature Links
FBLN7 PubMed Links

Gene Details

Gene
FBLN7
Gene Name
fibulin 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128165.1 Intron NP_001121637.1
NM_153214.2 Intron NP_694946.2
Gene
ZC3H8
Gene Name
zinc finger CCCH-type containing 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032494.2 Intron NP_115883.2

View Full Product Details