Product Details

SNP ID
rs61743157
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:30899330 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACACCCGAGGATCCGCCTTCAGTA[A/T]AGAATCGAAGAGAAACTGCTTCACT
Phenotype
MIM: 609534 MIM: 616422
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ATAD5 PubMed Links

Gene Details

Gene
ATAD5
Gene Name
ATPase family, AAA domain containing 5
There are no transcripts associated with this gene.

Gene
TEFM
Gene Name
transcription elongation factor, mitochondrial
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024683.3 1476 Missense Mutation ATA,TTA I308L NP_078959.3
XM_006722084.1 1476 Missense Mutation ATA,TTA I193L XP_006722147.1

View Full Product Details