Product Details

SNP ID
rs11544443
Assay Type
Functionally tested
NCBI dbSNP Submissions
37
Location
Chr.1:159782256 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTGGCTGCAGGAGATGCCATTGCTG[A/T]AATCCGACGACTACGACCCGGCTCC
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
DUSP23 PubMed Links

Gene Details

Gene
DUSP23
Gene Name
dual specificity phosphatase 23
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001319658.1 421 Missense Mutation GAA,GTA E124V NP_001306587.1
NM_001319659.1 421 Missense Mutation GAA,GTA E124V NP_001306588.1
NM_017823.4 421 Missense Mutation GAA,GTA E124V NP_060293.2

View Full Product Details