Product Details

SNP ID
rs35147534
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:31318369 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCTTTTCTTTCCCTTGTAGCTGT[A/G]AGCACTCCAGTAGATTGCATCACTA
Phenotype
MIM: 158380 MIM: 158381 MIM: 613113
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EVI2A PubMed Links

Gene Details

Gene
EVI2A
Gene Name
ecotropic viral integration site 2A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001003927.2 910 Silent Mutation CTC,CTT L238L NP_001003927.1
NM_014210.3 910 Silent Mutation CTC,CTT L215L NP_055025.2
Gene
EVI2B
Gene Name
ecotropic viral integration site 2B
There are no transcripts associated with this gene.

Gene
NF1
Gene Name
neurofibromin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000267.3 910 Intron NP_000258.1
NM_001042492.2 910 Intron NP_001035957.1
NM_001128147.2 910 Intron NP_001121619.1
XM_005257983.2 910 Intron XP_005258040.1
XM_005257984.2 910 Intron XP_005258041.1
XM_006721922.2 910 Intron XP_006721985.1
XM_006721924.2 910 Intron XP_006721987.1
XM_006721925.2 910 Intron XP_006721988.1
XM_006721926.3 910 Intron XP_006721989.1
XM_011524852.2 910 Intron XP_011523154.1
XM_011524857.2 910 Intron XP_011523159.1
XM_017024689.1 910 Intron XP_016880178.1
XM_017024690.1 910 Intron XP_016880179.1

View Full Product Details