Product Details

SNP ID
rs34198213
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:49446692 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCCAGCGACAGCCCCAGGGCTCC[A/T]TCCTGGAGAGGTGGCGGAATCAGGT
Phenotype
MIM: 613358 MIM: 611480 MIM: 605208
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ALDH16A1 PubMed Links

Gene Details

Gene
ALDH16A1
Gene Name
aldehyde dehydrogenase 16 family member A1
There are no transcripts associated with this gene.

Gene
PIH1D1
Gene Name
PIH1 domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017916.2 538 Silent Mutation GAA,GAT E230D NP_060386.1
XM_006723254.2 538 Silent Mutation GAA,GAT E103D XP_006723317.1
Gene
SLC17A7
Gene Name
solute carrier family 17 member 7
There are no transcripts associated with this gene.

View Full Product Details