Product Details

SNP ID
rs74818399
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:71549322 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAAATGGAATGAACCACTCCCTGC[C/G]CATTCCCTATAAGAATATCCCAAGA
Phenotype
MIM: 148021
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
KRTAP5-9 PubMed Links

Gene Details

Gene
KRTAP5-9
Gene Name
keratin associated protein 5-9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005553.3 903 UTR 3 NP_005544.4

View Full Product Details