Product Details

SNP ID
rs13014800
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:26792250 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCTTCAGCCGCCTGGTAAGCTCC[A/G]GGAGCTTCCCACCAACCCCTATGCC
Phenotype
MIM: 117139
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CENPA PubMed Links

Gene Details

Gene
CENPA
Gene Name
centromere protein A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042426.1 Intron NP_001035891.1
NM_001809.3 Intron NP_001800.1

View Full Product Details