Product Details
- SNP ID
-
rs8114118
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:45610188 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GAAGAATCCAGGGCTTCATGGTGCT[C/G]TCTGTGTGTATTTGGGTTAAGTTCT
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
WFDC9
PubMed Links
Gene Details
- Gene
- WFDC9
- Gene Name
- WAP four-disulfide core domain 9
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_147198.3 |
212 |
UTR 5 |
|
|
NP_671731.1 |
View Full Product Details