Product Details

SNP ID
rs61741503
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:87305784 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGGAGGCCGAGGGGCAGCAGACG[C/T]GCTGCAGGAGCCGGTAGCTGGTCCT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C16orf95 PubMed Links

Gene Details

Gene
C16orf95
Gene Name
chromosome 16 open reading frame 95
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195124.1 634 Silent Mutation GCA,GCG A212A NP_001182053.1
NM_001195125.1 634 Missense Mutation CAC,CGC H151R NP_001182054.1
NM_001256917.1 634 Missense Mutation CAC,CGC H136R NP_001243846.1

View Full Product Details