Product Details

SNP ID
rs4687994
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.3:119294711 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCACGGCGGCCCCGGAGCGGCGCGG[A/G]GTGGATCTCAGGCTCTGCCGGCCCG
Phenotype
MIM: 610911
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
ARHGAP31 PubMed Links

Gene Details

Gene
ARHGAP31
Gene Name
Rho GTPase activating protein 31
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020754.3 339 UTR 5 NP_065805.2
XM_006713714.3 339 UTR 5 XP_006713777.1
XM_017006955.1 339 UTR 5 XP_016862444.1

View Full Product Details