Product Details

SNP ID
rs149903512
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:87473010 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGGGCTGTCGGGGAACCATGGCT[G/T]CCCCGAGAGGTGAGAACTGGGTCTG
Phenotype
MIM: 605634
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CFAP206 PubMed Links

Gene Details

Gene
CFAP206
Gene Name
cilia and flagella associated protein 206
There are no transcripts associated with this gene.

Gene
SLC35A1
Gene Name
solute carrier family 35 member A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001168398.1 86 Missense Mutation GCC,TCC A3S NP_001161870.1
NM_006416.4 86 Missense Mutation GCC,TCC A3S NP_006407.1

View Full Product Details