Product Details

SNP ID
rs183995594
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:144466941 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACGTGACCCGAAATGTCTCCCGCC[C/G]TCCTCCCTGACCGGCAGCCAGCTCC
Phenotype
MIM: 616635 MIM: 603621 MIM: 615216
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CYHR1 PubMed Links

Gene Details

Gene
CYHR1
Gene Name
cysteine and histidine rich 1
There are no transcripts associated with this gene.

Gene
FOXH1
Gene Name
forkhead box H1
There are no transcripts associated with this gene.

Gene
KIFC2
Gene Name
kinesin family member C2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145754.3 Intron NP_665697.1
XM_005272357.3 Intron XP_005272414.1
XM_011517362.2 Intron XP_011515664.1

View Full Product Details