Product Details

SNP ID
rs564659491
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:60092120 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCAAGCCTGCTTTACGGCAGGGCC[C/G]GCCTCGGGAGCGAGCACAGACCGGG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PCNX4 PubMed Links

Gene Details

Gene
PCNX4
Gene Name
pecanex homolog 4 (Drosophila)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022495.5 845 UTR 5 NP_071940.4
XM_005268009.2 845 UTR 5 XP_005268066.1

View Full Product Details