Product Details

SNP ID
rs1037660073
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:104092846 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCCCACGTGAGCCATTTGCTCAC[C/T]CCTGTCTCTAACATCCCCTCAGCTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ASPG PubMed Links

Gene Details

Gene
ASPG
Gene Name
asparaginase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080464.2 Intron NP_001073933.2
Gene
LOC105370691
Gene Name
uncharacterized LOC105370691
There are no transcripts associated with this gene.

View Full Product Details