Product Details

SNP ID
rs7965126
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:121653265 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTCGTAGCGGTTCCCGTTCTCTGGG[A/G]GAAAGGACAGGGAGGTGTGGTGCAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MORN3 PubMed Links

Gene Details

Gene
MORN3
Gene Name
MORN repeat containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173855.4 Intron NP_776254.3
XM_005253869.4 Intron XP_005253926.1
XM_011538213.2 Intron XP_011536515.1
XM_011538214.2 Intron XP_011536516.1

View Full Product Details