Product Details

SNP ID
hCV25959520
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:41837827 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTTGGGTTGCCTGATAGACCCTAT[A/C]CAAAAGATGTAGTAGGGAAAAGGAG
Phenotype
MIM: 608778
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
KLHL10 PubMed Links

Gene Details

Gene
KLHL10
Gene Name
kelch like family member 10
There are no transcripts associated with this gene.

Gene
NT5C3B
Gene Name
5'-nucleotidase, cytosolic IIIB
There are no transcripts associated with this gene.

View Full Product Details