Product Details

SNP ID
rs9822259
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:38266130 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGCCTCATGTTCCGGCCACCCCC[C/T]GCCAATGCCAGCCTGCAGGACATCC
Phenotype
MIM: 604047
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC22A13 PubMed Links

Gene Details

Gene
SLC22A13
Gene Name
solute carrier family 22 member 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004256.3 225 Silent Mutation CCC,CCT P90P NP_004247.2
XM_017007519.1 225 UTR 5 XP_016863008.1

View Full Product Details