Product Details

SNP ID
rs34789013
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:2933651 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCTTTCCCACATTCAGTACATTC[C/G]TAGGGTTTGACCCCACTGTGTGATC
Phenotype
MIM: 194551
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ZNF77 PubMed Links

Gene Details

Gene
ZNF77
Gene Name
zinc finger protein 77
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021217.2 1632 Silent Mutation TAC,TAG Y492* NP_067040.1
XM_017027081.1 1632 Silent Mutation TAC,TAG Y312* XP_016882570.1
XM_017027082.1 1632 Silent Mutation TAC,TAG Y312* XP_016882571.1

View Full Product Details