Product Details

SNP ID
rs576651684
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:77801826 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTATTTATGTATTTTAAGTAAAATT[A/G]TAATCAGACATTTTGTAACCTGCTT
Phenotype
MIM: 616773
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIGA1 PubMed Links

Gene Details

Gene
MIGA1
Gene Name
mitoguardin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270384.1 Intron NP_001257313.1
NM_198549.3 Intron NP_940951.1
XM_005270846.4 Intron XP_005270903.1
XM_005270847.4 Intron XP_005270904.1
XM_005270848.4 Intron XP_005270905.1
XM_011541422.2 Intron XP_011539724.1
XM_017001227.1 Intron XP_016856716.1
XM_017001228.1 Intron XP_016856717.1
XM_017001229.1 Intron XP_016856718.1
XM_017001230.1 Intron XP_016856719.1
XM_017001231.1 Intron XP_016856720.1
XM_017001232.1 Intron XP_016856721.1

View Full Product Details