Product Details

SNP ID
rs759403335
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:50908368 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTTGTGAGTGGAAGAGGAAAAAA[A/C]CAGTCAAAATCTTCATTAATAGATA
Phenotype
MIM: 613741
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ABHD12B PubMed Links

Gene Details

Gene
ABHD12B
Gene Name
abhydrolase domain containing 12B
There are no transcripts associated with this gene.

Gene
PYGL
Gene Name
phosphorylase, glycogen, liver
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001163940.1 Intron NP_001157412.1
NM_002863.4 Intron NP_002854.3

View Full Product Details